Specifications
- Typ:Anti-UV, anti-scratch, anti-fog
- Färg:Klar/blå
- Beskrivning:SF201SGAF-BLU+ U6 lens marking
- Lins:Clear PC (2C-1.2 3M 1 FT)
- Förp.:1
Specifications
About this item
SecureFit™ 200 safety glass is lightweight, with 3M™ Pressure Diffusion Temple Technology. It feature a clear, anti-scratch and superior anti-fog lens, and a wraparound design that's secure and comfortable.
- Diffuse pressure over the ear with 3M™ Pressure Diffusion Temple Technology
- Soft, adjustable nose pads provide added comfort
- Flat and flexible temples combine easily with earmuffs and provide solid side protection
- Wraparound design gives the user a wide field of vision
- Scotchgard™ anti-fog coating offers superior anti-fog and anti-scratch properties
- Excellent protection against Ultraviolet (UV) radiation
- A removable foam-lined gasket is available
Wraparound, lightweight, and self-adjusting SecureFit™ 200 safety glass weigh just 18 g and offer a comfortable fit. They feature 3M™ Pressure Diffusion Temple Technology that allows the temple touchpoints to naturally adjust to an individual's head size. The clear lens features Scotchgard™ anti-fog coating to ensure clear visibility, resisting fogging longer than traditional anti-fog coatings even after washing multiple times, including using alcohol wipes or disinfecting by soaking in diluted bleach. Superior anti-fog and anti-scratch properties provide increased durability and visibility.
Wide, flexible temple arms over the ears provide solid side protection and work well with earmuffs. The wraparound design gives the user a wide field of vision and helps to reduce slippage from head movements.