Dysprosium ICP standards, (CRMs), Fluka™
Katalog # HONCE1018-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Koncentration:1000 mg/L
- Matris:2% HNO₃
- Förpackningstyp:Plastic bottle
- Förp.:100 mL
- Element:Dysprosium
- Symbol:Dy
Specifications
About this item
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).