Trace Metals III for AA and ICP-OES
Katalog # HONCE2168-100ML
Leverantör: Honeywell Chemicals
CAS-nummer:
Specifikationer
- Matris:2% HNO₃
- Förpackningstyp:Plastic bottle
- Förp.:100 mL
- Beskrivning:Trace Metals III
Specifikationer
Om denna produkt
Certified Reference Material (CRM).
500 mg/L each of Ba, Ca, Mo, Na, 100 mg/L each of K, Mg in HNO₃ 2%