Spike sample standard I for ICP
Katalog # HONCE2127-100ML
Leverantör: Honeywell Chemicals
CAS-nummer:
Specifikationer
- Matris:5% HNO₃
- Förpackningstyp:Plastic bottle
- Förp.:100 mL
- Beskrivning:Spike Sample Standard I
Specifikationer
Om denna produkt
Certified Reference Material (CRM).
200 mg/L each of Al, As, Ba, Se, Tl, 100 mg/L Fe, 50 mg/L each of Co, Mn, Ni, Pb, Sb, V, Zn, 25 mg/L Cu, 20 mg/L Cr, 5 mg/L each of Ag, Be, Cd in HNO₃ 5%