Order Entry
Sweden
ContactUsLinkComponent
Rhizopus (contains 50% Diatomaceous earth) Glucoamylase
Rhizopus (contains 50% Diatomaceous earth) Glucoamylase
Katalog # TCIAM0035-25G
Leverantör:  TCI
CAS-nummer:  
Rhizopus (contains 50% Diatomaceous earth) Glucoamylase
Katalog # TCIAM0035-25G
Leverantör:  TCI
CAS-nummer:  

Specifikationer

  • Förp.:
    25 g
  • Species:
    Rhizopus (contains 50% Diatomaceous earth)
  • Enzymnamn:
    Glucoamylase

Specifikationer

Om denna produkt