Pipe marker, H₂S
Pipe marker, H₂S, 150×12 mm
Katalog # BRDYN004308
Leverantör: Brady
Gå till:
Product Details & Documents
Individual pipe markers on map with perforated arrows, with pictogram.
- Rubber-based permanent adhesive
- Polyester with polyester overlaminate material
- Colour: Black on yellow
Specifikationer
- Beskrivning:Pipe marker, H₂S
- Mått:150×12 mm