Industrial gloves, oil repellent, HyFlex® 11-953
Supplier: Ansell
Certificates
About this item
These polyamide liners with rough nitrile palm coating provide protection against oil.
- Safe grip in oily and food environments
- Thin and flexible for outstanding comfort
- Excellent abrasion resistance
Recommended for: Assembly and finishing; assembly of white goods; maintenance; preparation of site - unloading of lines and valves; metal fabrication; general handling; quality control
Certifications: EN 388:2016 (4121A); Category II
Food contact approved
Food contact approved
Packaging: 12 pairs per bag.
Specifications
- Catalog No:
- ANSE11-953/9
- ANSE11-953/10
- ANSE11-953/6
- ANSE11-953/7
- ANSE11-953/8
- Modell:
- 11-953
- 11-953
- 11-953
- 11-953
- 11-953
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Polyamid
- Polyamid
- Polyamid
- Polyamid
- Polyamid
- Yta:
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Mätare:
- 13
- 13
- 13
- 13
- 13
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +