
Bariumklorid
Catalog # FFCHFF61120050
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:50 mL
- Seq:2370
Specifications
About this item
:
:208.24 g/mol
Merck Index:12,00998
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:∼963 °C
Density:2,33 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:233-788-1
UN:1564
ADR:6.1,III
:GHS06
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445