Bottles, Narrow Neck, Boston Round, PET, Rixius
Catalog # RIXI93100050028WC
Supplier: RIXIUS
New Product
Specifications
- Gänga:28/410
- Typ:Round
- Kapacitet:500 ml
- Färg:Klar
- Höjd:169 mm
- Material:PET
- Seq:02
Specifications
About this item
Round bottle made of PET. Can be used in combination with pump dispensers.
- Perfect alternative to glass
- Low dead weight, which means low transport and disposal costs
- Recyclable
- Almost unbreakable
- Food grade
Particularly suitable for homeopathic and cosmetic products, aromas, fragrance oils and dietary supplements.