AlphaTec® 37-220 Food Processing Gloves
Leverantör: Ansell
Certifikat
Om denna produkt
High performance 0,2 mm thick, nitrile reusable glove with UltraGrip™ technology.
- Lightweight reusable, nitrile gloves designed specifically for food processing and handling
- Ansell UltraGrip™ technology: thanks to the raised diamond-pattern surfacing it offers unmatched grip in wet and oily conditions
- Outstanding flexibility and dexterity, even at low temperature
- Liquid-proof gloves: Can be used on its own or as outer gloves over thermal liners or cut resistant gloves
- Latex-free formulation of these gloves presents no risk of Type 1 allergies
Recommended for: Dairy processing, fruit and vegetable processing, poultry processing, prepared meals.
Certifications: EN 388:2016 (2001X), EN ISO 374-1 Type B (JKT); EN ISO 374-5 VIRUS; Cat. III.
Packaging: 12 pairs per bag.
Specifikationer
- Catalog No:
- ANSE37-220/8
- ANSE37-220/10
- Modell:
- 37-220
- 37-220
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Latex/latexfri:
- Latexfria
- Latexfria
- Yta:
- Large diamond grip
- Large diamond grip
- Kragmodell:
- Kraghandske
- Kraghandske
- EN standard:
- EN 388:2016 (2001X), EN ISO 374-1 Type B (JKT); EN ISO 374-5 VIRUS
- EN 388:2016 (2001X), EN ISO 374-1 Type B (JKT); EN ISO 374-5 VIRUS
- Godkända för livsmedelskontakt:
- +
- +