5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin ≥95.0%
Catalog # TCIAT1497-100MG
Supplier: TCI
CAS Number:
Specifications
- Förp.:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (HPLC):min. 95.0 area%
- Lambda max.:416.0 to 420.0 nm (MeOH)
- Molar extinction coefficient:min. 400000 (MeOH)
Specification Test Results
About this item
:
:678.75 g/mol
CAS Number:51094-17-8
Formula:C₄₄H₃₀N₄O₄
MDL Number:MFCD00191501