Triphenylborane sodium hydroxide adduct 7 - 10% i vatten
Catalog # TCIAT1085-500G
Supplier: TCI
CAS Number:
Specifications
- Förp.:500 g
- Seq:3645
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Concentration (Neutralization titration):7.0 to 10.0 w/w%
Specification Test Results
About this item
:
:282.12 g/mol
EINECS:235-171-2
UN:1760
ADR:8,III
:GHS05
:GHS07
CAS Number:12113-07-4
Formula:C₁₈H₁₅B
MDL Number:MFCD00003008