Kiselgel, spherical 100 µm for column chromatography
Catalog # TCIAS0822-1KG
Supplier: TCI
CAS Number:
Specifications
- Förp.:1 kg
- Seq:4024
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Specific surface:gt. 400 to 550 m2/g
- Pore volume:0.70 to 0.90 mL/g
- Density of arrangement:0.40 to 0.60 g/mL
- Water content:3.0 to 7.0 %
Specification Test Results
About this item
:
:60.08 g/mol
Merck Index:14,08493
EINECS:231-545-4
:GHS07
CAS Number:7631-86-9
:112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232