Order Entry
Sweden
ContactUsLinkComponent
Kiselgel, spherical 60 µm för flashkromatografi
Kiselgel, spherical 60 µm för flashkromatografi
Catalog # TCIAS0821-1KG
Supplier:  TCI
CAS Number:  
undefined
Kiselgel, spherical 60 µm för flashkromatografi
Catalog # TCIAS0821-1KG
Supplier:  TCI
CAS Number:  

Specifications

  • Förp.:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Specific surface:
    400 to 600 m2/g
  • Pore volume:
    0.70 to 0.90 mL/g
  • Density of arrangement:
    0.40 to 0.60 g/mL
  • Water content:
    3.0 to 7.0 %

Specification Test Results

About this item

 :
 :60.08 g/mol
Merck Index:14,08493
EINECS:231-545-4
 :GHS07
CAS Number:7631-86-9
 :112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232