Sodium tris(1,1,1,3,3,3-hexafluoroisopropoxy)borohydride ≥75.0% reducing reagent
Catalog # TCIAS0810-5G
Supplier: TCI
CAS Number:
Specifications
- Förp.:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 75.0 %
Specification Test Results
About this item
:
:535.90 g/mol
Storage Temperature:Kylskåp
UN:1409
ADR:4.3,II
:GHS02
CAS Number:139494-68-1
Formula:C₉H₄BF₁₈NaO₃