Olje rött O for biochemical research
Catalog # TCIAO0483-25G
Supplier: TCI
CAS Number:
Specifications
- Förp.:25 g/mol
- Seq:9230
Specifications
Specification Test Results
- Lambda max.:510.0 to 520.0 nm (2-Propanol)
- Cell staining test:to pass test
Specification Test Results
About this item
:
:408.51 g/mol
CAS Number:1320-06-5
Formula:C₂₆H₂₄N₄O
MDL Number:MFCD00003898