4-Nitrobenzenediazonium tetrafluoroborate ≥98.0% (by HPLC, titration analysis)
TCIAN0137-25G
: TCI
:
- Förp.:25 g
- Seq:3308
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
:
:236.92 g/mol
:Frys
:207-261-1
:1759
:8,III
:GHS05
:GHS07
:456-27-9
:C₆H₄BF₄N₃O₂
:MFCD00012005