Order Entry
Sweden
ContactUsLinkComponent
Potassium-7-hydroxy-1-naphthalenesulphonate ≥90.0% (contains isomer)
Potassium-7-hydroxy-1-naphthalenesulphonate ≥90.0% (contains isomer)
Catalog # TCIAN0075-25G
Supplier:  TCI
CAS Number:  
Potassium-7-hydroxy-1-naphthalenesulphonate ≥90.0% (contains isomer)
Catalog # TCIAN0075-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Förp.:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • Drying loss:
    max. 3.0 %

Specification Test Results

About this item

 :
 :262.32 g/mol
EINECS:250-101-0
 :GHS07
CAS Number:30252-40-5
Formula:C₁₀H₈O₄S
MDL Number:MFCD00065327