2-Methoxyphenothiazine ≥98.0%
Catalog # TCIAM0822-25G
Supplier: TCI
CAS Number:
Specifications
- Förp.:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light gray to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
Specification Test Results
About this item
:
:229.30 g/mol
EINECS:217-195-5
CAS Number:1771-18-2
Formula:C₁₃H₁₁NOS
MDL Number:MFCD00037838