5-Methylisoxazole ≥95.0%
Supplier: TCI
Certificates
About this item
CAS Number:5765-44-6
:
Formula:C₄H₅NO
MDL Number:MFCD00003152
:83.09 g/mol
Boiling Pt:122°C
EINECS:227-289-8
UN:1993
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %