Order Entry
Sweden
ContactUsLinkComponent
Harmine ≥98.0%
Harmine ≥98.0%
Catalog # TCIAH0001-1G
Supplier:  TCI
CAS Number:  
Harmine ≥98.0%
Catalog # TCIAH0001-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Förp.:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :212.25 g/mol
Merck Index:14,04616
EINECS:207-131-4
 :GHS07
 :GHS08
CAS Number:442-51-3
Formula:C₁₃H₁₂N₂O
MDL Number:MFCD00004958