
(+)-Benzylmethylphenylsilylacetic acid ≥98.0% for e.e. determination by NMR
TCIAB1720-100MG
: TCI
:
- Förp.:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Optical purity (LC):min. 99.8 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:68.0 to 72.0 deg-C
- Specific rotation [a]20/D:+5.0 to +8.0 deg (C=2, CHCl3)
:
:270.40 g/mol
:GHS07
:95373-54-9
:C₁₆H₁₈O₂Si