
Nonafluorovaleric acid ∼0.5 mol/L i vatten för jonparkromatografi, for LC-MS
TCIAA5714-10ML
: TCI
:
- Förp.:10 mL
- Seq:2275
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):0.50 to 0.55 mol/L
- Absorbance (270nm):max. 0.300
- Absorbance (300nm):max. 0.100
:
:8,II
:264.05 g/mol
:220-300-7
:3265
:GHS05
:2706-90-3
:C₅HF₉O₂
:MFCD00040211