Triacetin, Millipore®
Catalog # 1.08238.0500
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:500 ml
- Seq:2725
Specifications
About this item
:
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1,1596 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:138 °C (closed cup)
Storage Temperature:Omgivande
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716