Triacetin
Catalog # EHERC17608500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:0,25 g
- Seq:3029
Specifications
About this item
:
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1,1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Omgivande
EINECS:203-051-9
CAS Number:102-76-1
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716