Acetaldehyde ∼2% i DMF for detection of primary and secondary amines
Catalog # TCIAA1640-10ML
Supplier: TCI
CAS Number:
Specifications
- Förp.:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Concentration (Oxime Formation):2.0 to 2.3 w/w%
Specification Test Results
About this item
:
:44.05 g/mol
Merck Index:14,00039
Storage Temperature:Kylskåp
EINECS:200-836-8
UN:1992
ADR:3,III
:GHS02
:GHS05
:GHS06
:GHS08
CAS Number:75-07-0
Formula:CH₃CHO
MDL Number:MFCD00006991