Petrochemical filter stick apparatus with hooks and springs
Katalog # SCERSP513880-0000
Leverantör: DWK Life Sciences
Specifikationer
- Beskrivning:Petrochemical filter stick apparatus
- Förp.:1
Specifikationer
Om denna produkt
The Filter Stick Apparatus is used in the determination of oil content and solvent extractables in petroleum waxes.
- Supplied with one pair of springs
- Designed from ASTM D721 specifications
- Manufactured from borosilicate 3,3 glass
Assembly consists of a sintered glass filter stick with air pressure inlet tube and delivery nozzle and a cooling tube with NS 24/40 joint.
Certifications: The borosilicate glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.