L-Homoserine för syntes, Sigma-Aldrich®
Catalog # 8.14359.0001
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:1 g
- Seq:3047
Specifications
Specification Test Results
- Solubility:1100 g/l (30 °C)
- Molar Mass:119.12 g/mol
- Hill Formula:C4H9NO3
Specification Test Results
About this item
:
:119.12 g/mol
Merck Index:13,04760
Melting Pt:203 °C
:M-Clarity™ quality level = MQ200
EINECS:211-590-6
CAS Number:672-15-1
Formula:C₄H₉NO₃
MDL Number:MFCD00063090