Fosforwolframsyra, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:12501-23-4
:12067-99-1
Formula:H₃[P(W₃O₁₀)₄]·nH₂O
MDL Number:MFCD00011342
:2898.07 g/mol
Merck Index:13,07450
Melting Pt:107 °C
Storage Temperature:Omgivande
EINECS:215-682-7
UN:3260
ADR:8,III
:M-Clarity™ quality level = MQ300
:GHS05
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:107 °C
- Molar Mass:2880.17 g/mol (anhydrous)
- Bulk Density:960 kg/m3
- pH:1.3 (20 g/l, H2O, 20 °C)