Instant Buffer K40 pH4.0 (acc. to Kolthoff)
Katalog # TCIAI0234-1SET
Leverantör: TCI
CAS-nummer:
Specifikationer
- Förp.:1 SET
- Pk Info:50 packs
- pH-värde:4.0
Specifikationer
Om denna produkt
Form: White granules
Buffer prepared from a mixture of succinic acid and borax according to Kolthoff.