Orders LinkContactUsLinkComponent
VWR®, Dot Printer for VWR® Balances
VWR®, Dot Printer for VWR® Balances
TX-110 dot printer for balances
Katalog # 611-4790
Leverantör:   VWR Collection
VWR®, Dot Printer for VWR® Balances
Katalog # 611-4790
Leverantör:  VWR Collection
VWR®, Dot Printer for VWR® Balances
VWR®, Dot Printer for VWR® Balances
TX-110 dot printer for balances
Price
6 275,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
The TX-110 provides high speed dot printing and features a 184×64 OLED display.
  • Printouts are suitable for long-term preservation
  • ISO/GLP compliant data printout
  • Built-in real-time clock
  • Sample name and batch number can be input and printed
  • Sum, mean and formulation function
Specifikationer
  • Beskrivning:
    TX-110 dot printer for balances
  • Seq:
    001
Accessories
Produktinformation
  • Katalog #
    För
    Tillgänglighet
    Price
  • Paper roll for dot printer
    Paper roll for dot printer
    611-4791
    TX-110 printer (611-4790)
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 170,00 kr