Order Entry
Sweden
ContactUsLinkComponent
Etylacetat ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Etylacetat ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65812-200L
   Safer Human and Environmental Health
CAS Number:  
Etylacetat ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65812-200L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Förpackningstyp:
    Metal drum
  • Förp.:
    200 L
  • Seq:
    5549
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Miljöcertificering:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    76 - 78 °C
  • Density (D 20/4):
    0.899 - 0.901
  • Refractive index (n 20/D):
    1.3710 - 1.3730
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.025 %
  • Free acid (as CH₃COOH):
    Max. 0.0045 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Bismuth (Bi):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Potassium (K):
    Max. 0.00005 %
  • Lithium (Li):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Molybdenum (Mo):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Sodium (Na):
    Max. 0.00005 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Strontium (Sr):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • methyl acetate:
    Max. 0.1 %
  • Ethanol (GC):
    Max. 0.1 %
  • Isobutyl alcohol (GC):
    Max. 0.1 %
  • Methanol (GC):
    Max. 0.1 %
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10

Specification Test Results

About this item

 :
 :88.11 g/mol
Boiling Pt:77 °C
Density:0,901 g/cm3
Flash Pt:–4 °C
UN:1173
ADR:3,II
 :GHS07
 :GHS02
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171