VWR®, Wire basket with handle
Catalog # 216-2254
Supplier: VWR Collection
Specifications
- B×D×H:400×300×200 mm
- Förp.:1
Specifications
About this item
This wire basket is made from rust-proof, electrolytically polished stainless steel.
- Very good chemical and temperature resistance
- Foldaway carrying handle with plastic hand grip
- Robust design with two rods for base reinforcement
Suitable for transportation or sterilisation of laboratory goods.