Chemical resistant gloves, PVC, Showa 650
: Showa
The 650 chemical resistant glove offers anti-odour and antibacterial protection as well as a moisture-wicking soft cotton liner.
- Full PVC coating and extra rough-finish PVC over the entire hand protects against chemicals, grease and liquid
- Anatomical hand shape reduces hand fatigue
- Seamless knit prevents irritation during long periods of wear
- Ultra-supple and waterproof glove with a rough finish
- Impermeable for working in damp or greasy environments
- Comfortable and soft, allows objects to be gripped firmly
Suitable applications: Chemical spray and treatment, coating preparation, painting & spray workshops.
: EN 388:2016 (4121X); EN 374-1:2016 Type B (JKL); EN 374-5:2016; Cat. II
: 10 pairs per polybag.
- Catalog No:
- 112-5872
- 112-5871
- 112-5874
- 112-5873
- Modell:
- Showa 650
- Showa 650
- Showa 650
- Showa 650
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Bomull
- Bomull
- Bomull
- Bomull
- Yta:
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Full PVC coating
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (4121X); EN 374-1:2016 Type B (JKL); EN 374-5:2016
- EN 388:2016 (4121X); EN 374-1:2016 Type B (JKL); EN 374-5:2016
- EN 388:2016 (4121X); EN 374-1:2016 Type B (JKL); EN 374-5:2016
- EN 388:2016 (4121X); EN 374-1:2016 Type B (JKL); EN 374-5:2016