Stereo microscopes, ergonomic, Mantis
Catalog # VISIMEO-010
Supplier: VISION ENGINEERING
Specifications
- Beskrivning:MEO objective 10× for Mantis Elite
- Förp.:1
Specifications
About this item
Superior imaging for a wide range of inspection and rework tasks.
- Patented optical technology for fatigue-free viewing and superb image quality
- Wide range of magnification options to 20x
- Long working distances; large depths of field
- Shadow-free true colour LED illumination
- Choice of table stand or articulated arm
Mantis Compact excels in the low magnification range for inspection or manipulation tasks where bench magnifiers have traditionally been used. Mantis Compact has a small footprint and low nvestment cost giving a superb price/performance ratio.
Mantis Elite has enhanced optical performance, including higher magnification, a large field of view plus long working distances, making it a perfect alternative to traditional stereo microscopes for a wide range of inspection, preparation and manipulation tasks requiring hand-eye co-ordination.
Ordering information: Objective lenses sold separately.