TISAB I Solution, Supelco®
Katalog # 89465-500ML
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förpackningstyp:Plastic bottle
- Förp.:500 mL
- Beskrivning:TISAB I solution
- M-Clarity quality level:MQ100
Specifikationer
Om denna produkt
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes.
TISAB I can be prepared using NaCl with glacial acetic acid, tri-sodium citrate and NaOH.
TISAB I has been used in determination of fluoride-ion concentration using fluoride ion sensitive electrode.