Single-use temperature logger tempmate.®-S1 V3
Product Details & Documents
This third generation of single-use disposable logger is for temperature recording. Options include up to six selectable temperature limits (high and low) for even more detailed analysis, a unique 'start - alarm - delay' in order to avoid a premature alert, and a time-scheduled start that makes internal handling easy. No recalibration necessary.
- Pre-configured
- High precision digital sensor
- Extended run time (up to 200 days)
- Double-memory (31200 measuring points)
- Increased accuracy (average accuracy of ±0,2 °C)
- External sensor for improved response time (T90)
No IT expenses, as there is no software. No employee software training needed. Familiar simple handling remains unchanged. Guided and secure USB. Improved button response provide simple operation.
Data security via two separate flash memories. PDF and CSV file is non manipulable. Password protection possible. More stable circuit board by changing from THT to SMD assembly. Special lamination protects the circuit board against external influences. Additional protection of the hardware through new and reinforced housing. PDF and CSV reports are available at any time without stopping the logger.
Use of recycled plastics. Raw materials are preserved through digitalisation (no more hard copies of Shipping Documents and Validation Certificates, these documents are available online). No more zip bags required. Environmentally friendly packaging.
Shelf life approx. 24 months.
Certifications: According to EN 12830 and FCC, CE, RoHS. Validation certificate (6 points as PDF in memory for each appliance). Every logger has it own certificate saved on the logger itself.
Beställningsinformation: Various models with pre-configured alarm and delay settings are available. For models with other configurations, contact your Avantor representative.