Order Entry
Sweden
ContactUsLinkComponent
 
Flammable
Health
Irritant
Petroleumeter, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Catalog # 261734-1L
CAS Number:  
Flammable
Health
Irritant
Petroleumeter, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Catalog # 261734-1L
CAS Number:  

Specifications

  • Förpackningstyp:
    Glass bottle
  • Förp.:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Explosion Limit:
    8 %
  • Identity (IR):
    complying
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 260 nm:
    Max. 0.01
  • Absorbance at 240 nm:
    Max. 0.05
  • Absorbance at 230 nm:
    Max. 0.10
  • Absorbance at 220 nm:
    Max. 0.30
  • Absorbance at 210 nm:
    Max. 1.0
  • Vapor Density:
    2.5 (vs air)
  • Refractive Index:
    n20/D 1.363 (lit.)

Specification Test Results

About this item

 :
Boiling Pt:40…60 °C (1,013 hPa)
Density:0,65 g/cm³ (20 °C)
Flash Pt:–30 °C
Storage Temperature:Omgivande
UN:3295
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849