Avantor® Hichrom PEEKsil™, Tubing
Catalog # HICHHI-1083
Supplier: Avantor
Specifications
- Beskrivning:PEEKsil™ tubing for HPLC, LC/MS applications
- Längd:500 mm
- Seq:02
- Förp.:2
Specifications
About this item
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.