VWR®, Bag Rack for Blender Bags
Catalog # 129-1474
Supplier: VWR Collection
Specifications
- Fack:10
- Beskrivning:Bag rack, stainless steel
- Förp.:1
Specifications
About this item
This bag rack is used for batch preparation of samples prior to blending as well as sample storage after blending.
- Bags are temporarily sealed using closure clips and are then suspended in the bag rack support slots
- It can store up to 10, 400 series blender bags
The rubber feet keep it very stable on the counter top and the bottom basin can easily hold 400 ml of liquid in case of a mishap. The bag rack fits in most incubators and is autoclavable as well.