Sleeve protectors, PP
Katalog # HOIT109760
Leverantör: HUBEI ORIENT INTL TRADING
Specifikationer
- Outfit applikationer:Laboratory,Food
- Färg:Vit
- Längd:475 mm
- Förp.:400
Specifikationer
Om denna produkt
These disposable sleeve protectors made of PP provide waterproof protection for light-duty tasks.
- Lightweight and soft
- Waterproof and oil-proof
- Breathable fabric
- Latex-free
Ideal for use in non-hazardous applications, such as cleaning, maintenance, food processing and handling, manufacturing, hotels and catering.
Weight - 40 g/m²