Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
SYBR® Green II
SYBR® Green II nucleic acid stain
SYBR® Green II nucleic acid stain
SYBR® Green II nucleic acid stain
Catalog # 730-1294
Supplier:
LONZA
CAS Number:
Expand image
SYBR® Green II nucleic acid stain
Catalog # 730-1294
Supplier:
LONZA
CAS Number:
Specifications
Förp.:
500 µl
Seq:
1418
Pk Info:
10× 50 μl
Specifications
About this item
Product Details
:
CAS Number:
172827-25-7
MDL Number:
MFCD00284716
Product Details