Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Standarder
Organic Standards
N-Nitrosoanatabine, analytical standard, Supelco®
N-Nitrosoanatabine, analytical standard, Supelco®
N-Nitrosoanatabine, analytical standard, Supelco®
Catalog # 75281-5MG
Supplier:
Merck
CAS Number:
Expand image
N-Nitrosoanatabine, analytical standard, Supelco®
Catalog # 75281-5MG
Supplier:
Merck
CAS Number:
Specifications
Förp.:
5 mg
Standard:
analytical standard
Beskrivning:
N-Nitrosoanatabine
Benämning:
N-Nitrosoanatabine
Specifications
About this item
Product Details
Product Details