PAS-färgning, Sigma-Aldrich®
Catalog # 1.01646.0001
Supplier: Merck
CAS Number:
Specifications
- Förp.:400 Tests
- Seq:0810
- Pk Info:2 x 500 ml (Preparation Capacity 2500 - 3000 slides)
Specifications
Specification Test Results
- Suitability for microscopy:passes test
- nuclei:Blue
- acid mucosubstances:light blue
- polysaccharides:purple
- neutral mucopolysaccharides:purple
Specification Test Results
About this item
:
Storage Temperature:Omgivande