ContactUsLinkComponent
Laboratory blender, Seward Stomacher® 4500
Laboratory blender, Seward Stomacher® 4500
Katalog # SEWA4500/000/EU
Leverantör:  Seward (FermionX)
undefined
Laboratory blender, Seward Stomacher® 4500
Katalog # SEWA4500/000/EU
Leverantör:  Seward (FermionX)
Specifikationer
  • Beskrivning:
    Laboratory paddle blender, Stomacher® 4500 BAM
  • Modell:
    Stomacher® 4500
  • Vikt:
    43,5 kg
  • B×D×H:
    610×610×584 mm
  • Förp.:
    1
Specifikationer
Om denna produkt
The robust Stomacher® 4500 is ideal for the BAM large volume testing procedures from 1000 to 4500 ml.
  • Easy to clean
  • Easy bag loading
  • Programmable settings
  • 3 year warranty, 10 year service and spare parts
Stomacher® lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples. The Stomacher® machines work by the action of twin-reciprocating paddles on the sample contained inside a sterile plastic bag.
Expanding the food testing focused blenders, the Stomacher® 4500 BAM is the perfect tool for larger volume BAM testing procedures for extraction from 375 g composite samples.