Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, Urethane, analytical standard, Supelco®
Organic reference standard, Urethane, analytical standard, Supelco®
Katalog # 76607-100MG
Leverantör:  Merck
CAS-nummer:  
undefined
Organic reference standard, Urethane, analytical standard, Supelco®
Katalog # 76607-100MG
Leverantör:  Merck
CAS-nummer:  

Specifikationer

  • Förp.:
    100 mg
  • Standard:
    Analytical standard
  • CAS-nr:
    51-79-6
  • Beskrivning:
    Urethane, ≥98,0% (GC)
  • Smältpunkt:
    46...49 °C
  • Formel:
    C₃H₇NO₂ 
  • Benämning:
    Urethane
  • Kokpunkt:
    182...184 °C (lit.)
  • Form:
    Neat

Specifikationer

Om denna produkt

Urethane is used as an anesthetic in animals, since it shows minimal effects on cardiovascular and respiratory systems and maintenance of spinal reflexes.

It can also be used in the synthesis of poly(urethane urea) segmented block, which can find applications as blood sacs in ventricular assist devices and total artificial hearts.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.