Organic reference standard, Triethanolamine, analytical standard, Supelco®
Katalog # 50926-1ML
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:102-71-6
- Beskrivning:Triethanolamine, ≥99,0% (GC)
- Smältpunkt:17,9...21 °C (lit.)
- Formel:C₆H₁₅NO₃
- Benämning:Triethanolamine
- Kokpunkt:190...193 °C/5 mm Hg (lit.)
- Form:Neat