Organic reference standard, Pentyl acetate, analytical standard, Supelco®
66962-1ML
: Merck
:
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:628-63-7
- Beskrivning:Pentyl acetate, ≥97.0% (GC)
- Smältpunkt:–100 °C
- Formel:C₇H₁₄O₂
- Benämning:Pentyl acetate
- Flampunkt:39 °C
- Kokpunkt:142...149 °C
- Molecular weight (Mw):C₇H₁₄O₂
- Form:Neat
Used as banana essence and test odorant in studies of olfactory function and in studies of the psychosocial effects of odor.