Organic reference standard, Methyl dihydrojasmonate, mixture of cis and trans, analytical standard, Supelco®
59677-1ML
: Merck
:
- Förp.:1 mL
- Standard:Analytical standard
- CAS-nr:24851-98-7
- Beskrivning:Methyl dihydrojasmonate, mixture of cis and trans, ≥96,0% (GC)
- Formel:C₁₃H₂₂O₃
- Benämning:Methyl dihydrojasmonate
- Flampunkt:113 °C
- Kokpunkt:110 °C/0,2 mmHg