Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, Isoborneol, analytical standard, Supelco®
Organic reference standard, Isoborneol, analytical standard, Supelco®
Catalog # 79856-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Isoborneol, analytical standard, Supelco®
Catalog # 79856-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Förp.:
    100 mg
  • Standard:
    Analytical standard
  • CAS-nr:
    124-76-5
  • Beskrivning:
    Isoborneol, ≥95,0% (GC)
  • Smältpunkt:
    212 - 214 °C
  • Formel:
    C₁₀H₁₈O
  • Benämning:
    Isoborneol

Specifications

About this item

Isoborneol is a monoterpene and a known antioxidant, which is a component of several plant essential oils.